Learn More
Aviva Systems Biology Corp NRF1 Antibody - C-terminal region (ARP38543_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38543_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NRF1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human NRF1. Alternate names: ALPHA-PAL, EWGApplicationIHC, WB. Species reactivity: Human, Rat
Catalog No. 50-014-56696