Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nrf2 (69-84) | 1364173-77-2 | 95.25% | 1858.01 | 25 MG
SDP

Catalog No. 5000426961
Encompass_Preferred

Nrf2 (69-84) is a peptide fragment of Nrf2 protein that contains the key ETGE motif, which is an important region for binding to the Kelch domain of Keap1 protein. It can be used to study the role of Nrf2 in the development and progression of diseases, especially in cancer, neurodegenerative diseases, and inflammatory diseases.

  • Peptide fragment of Nrf2 protein
  • Contains key ETGE motif for binding to Keap1 protein
  • Suitable for studying Nrf2's role in cancer
  • Useful for research into neurodegenerative diseases
  • Applicable for inflammatory diseases research
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • Soluble in H2O at 9.09 mg/mL (4.89 mM)

Catalog No. 50-004-26961 Supplier Medchemexpress LLC Supplier No. 25MGHYP10530
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nrf2 (69-84) is a peptide fragment of Nrf2 protein that contains the key ETGE motif, which is an important region for binding to the Kelch domain of Keap1 protein. It can be used to study the role of Nrf2 in the development and progression of diseases, especially in cancer, neurodegenerative diseases, and inflammatory diseases.

  • Peptide fragment of Nrf2 protein
  • Contains key ETGE motif for binding to Keap1 protein
  • Suitable for studying Nrf2's role in cancer
  • Useful for research into neurodegenerative diseases
  • Applicable for inflammatory diseases research
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • Soluble in H2O at 9.09 mg/mL (4.89 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.