Learn More
Medchemexpress LLC Nrf2 (69-84) | 1364173-77-2 | 95.3% | 1858.01 | 5 MG
SDP
Catalog No. 5000425128
Encompass_Preferred
Nrf2 (69-84) is a peptide fragment of the Nrf2 protein, featuring the critical ETGE motif. This key region binds to the Kelch domain of the Keap1 protein. It is utilized in research to explore Nrf2's role in the development and progression of various diseases.
  • Peptide fragment of Nrf2 protein
  • Contains critical ETGE motif
  • Binds to Kelch domain of Keap1 protein
  • Used for research on Nrf2 in disease development
  • Relevant for cancer, neurodegenerative, and inflammatory disease studies
Catalog No. 50-004-25128 Supplier Medchemexpress LLC Supplier No. HYP105305MG
Nrf2 (69-84) is a peptide fragment of the Nrf2 protein, featuring the critical ETGE motif. This key region binds to the Kelch domain of the Keap1 protein. It is utilized in research to explore Nrf2's role in the development and progression of various diseases.
  • Peptide fragment of Nrf2 protein
  • Contains critical ETGE motif
  • Binds to Kelch domain of Keap1 protein
  • Used for research on Nrf2 in disease development
  • Relevant for cancer, neurodegenerative, and inflammatory disease studies