Learn More
Medchemexpress LLC Nrf2 activator-12 | 3020661-05-3 | 98.6% | 313.20 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16920110MM/1ML
Encompass_Preferred
Nrf2 activator-12 (compound 10v) is a potent activator of Nrf2 with an EC50 of 83.5 nM. It demonstrates promising pharmacological properties, including reversing disease progression and alleviating demyelination in an experimental autoimmune encephalomyelitis mouse model.
  • Potently activates Nrf2 (EC50=83.5 nM).
  • Exhibits promising pharmacological properties.
  • Reverses disease progression in an experimental autoimmune encephalomyelitis mouse model.
  • Alleviates demyelination in an experimental autoimmune encephalomyelitis mouse model.
Catalog No. 50-003-90911