Learn More
Genetex Inc Anti-NRF2 antibody [N2C2], Internal
SDP
Supplier:  Genetex Inc GTX103322
Encompass
The Anti-NRF2 antibody [N2C2], Internal from GeneTex is a Rabbit Polyclonal antibody to NRF2. The Anti-NRF2 antibody [N2C2], Internal Recognizes Human, Mouse, Rat, Bird. The Anti-NRF2 antibody [N2C2], Internal has been validated for the following Applications: IP, ICC/IF, ChIP assay, IHC, WB, IHC-P.
Catalog No. NC1167792