Learn More
Medchemexpress LLC HY-101025 100mg Medchemexpress, Nrf2-IN-1 CAS:1610022-76-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101025100MG
Encompass_Preferred
Medchemexpress, HY-101025 100mg Nrf2-IN-1 CAS:1610022-76-8 Nrf2-IN-1 (Compound 4f) is an inhibitor of nuclear factor-erythroid 2-related factor 2 (Nrf2) , acts as a promising agent in acute myeloid leukemia (AML) therapy [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2011