Learn More
TARGETMOL CHEMICALS INC NRF2-IN-1 5MG
SDP
Catalog No. 502581474
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nrf2-IN-1 is an nuclear factor-erythroid 2-related factor 2 (Nrf2) inhibitor. Nrf2-IN-1 is developed for the research of acute myeloid leukemia (AML). purity: 99%
Catalog No. 50-258-1474 Supplier TARGETMOL CHEMICALS INC Supplier No. T122535MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nrf2-IN-1 is an nuclear factor-erythroid 2-related factor 2 (Nrf2) inhibitor. Nrf2-IN-1 is developed for the research of acute myeloid leukemia (AML). purity: 99%