Learn More
Enzo Life Sciences Nrf 2 polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPTF125D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide derived from the sequence of human Nrf2 (residues 40-60) with a cysteine residue added and coupled to KLH. UniProt: Q16236. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Hamster, Guinea pig, Dog. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-666-629