Learn More
Abeomics Nrf2 Reporter Mcf7 Cell Line
Catalog No. NC1745332
Encompass_Preferred
The Nrf2 Leeporter Luciferase Reporter cell line is a stably transfected MCF7 cell line which expresses Renilla luciferase reporter gene under the transcriptional control of the antioxidant response element (ARE). ARE is known to regulate expression and induction of various detoxifying enzyme genes in response to antioxidants and xenobiotics, and is primarily regulated by the Keap1-Nrf2 pathway in which induction and nuclear translocation of Nrf2 mediated by antioxidants and xenobiotics results in the binding of Nrf2 to ARE leading to the expression of defensive genes. Cell line application: (1) Monitor the Nrf2 induction activity, and (2) Screen for activators or inhibitors of the Nrf2 signaling pathway. .
Catalog No. NC1745332 Supplier Abeomics Supplier No. 14117ACL
The Nrf2 Leeporter Luciferase Reporter cell line is a stably transfected MCF7 cell line which expresses Renilla luciferase reporter gene under the transcriptional control of the antioxidant response element (ARE). ARE is known to regulate expression and induction of various detoxifying enzyme genes in response to antioxidants and xenobiotics, and is primarily regulated by the Keap1-Nrf2 pathway in which induction and nuclear translocation of Nrf2 mediated by antioxidants and xenobiotics results in the binding of Nrf2 to ARE leading to the expression of defensive genes. Cell line application: (1) Monitor the Nrf2 induction activity, and (2) Screen for activators or inhibitors of the Nrf2 signaling pathway. .