Learn More
Cayman Chemical Nrf2 Transcription Factor Assay Kit, 96 Well
SDP
Catalog No. 502894376
Encompass_Preferred
Nuclear factor E2-related factor 2 (Nrf2) is a transcription factor that plays a key role in maintaining redox homeostasis via its interaction with a cysteine-rich protein Kelch-like ECH-associated protein 1 (Keap1). In resting cells, Nrf2 and Keap1 form a tight complex, which is targeted for degradation by proteasomes. Under oxidative stress, Nrf2 is released from the Nrf2/Keap1 complex and translocates to the nucleus where it is able to induce the expression of a battery of genes encoding diverse cytoprotective proteins, including antioxidative enzymes, anti-inflammatory mediators, and proteasomes. Cayman’s Nrf2 Transcription Factor Assay is a non-radioactive, colorimetric method for detecting specific transcription factor DNA binding activity in nuclear extracts. A specific double stranded DNA (dsDNA) sequence containing the Nrf2 response element is immobilized onto the wells of a 96-well plate. Nrf2 contained in nuclear.
Catalog No. 50-289-4376 Supplier Cayman Chemical Supplier No. 60059096 WELL
Nuclear factor E2-related factor 2 (Nrf2) is a transcription factor that plays a key role in maintaining redox homeostasis via its interaction with a cysteine-rich protein Kelch-like ECH-associated protein 1 (Keap1). In resting cells, Nrf2 and Keap1 form a tight complex, which is targeted for degradation by proteasomes. Under oxidative stress, Nrf2 is released from the Nrf2/Keap1 complex and translocates to the nucleus where it is able to induce the expression of a battery of genes encoding diverse cytoprotective proteins, including antioxidative enzymes, anti-inflammatory mediators, and proteasomes. Cayman’s Nrf2 Transcription Factor Assay is a non-radioactive, colorimetric method for detecting specific transcription factor DNA binding activity in nuclear extracts. A specific double stranded DNA (dsDNA) sequence containing the Nrf2 response element is immobilized onto the wells of a 96-well plate. Nrf2 contained in nuclear.