Learn More
Yamato Scientific America, Inc. Yamato 286L Refrigerant Incubator, Eco-friendly, 115V/220V
Catalog No. 501898108 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Programmable , eco-friendly 286L refrigerant incubator. Upgraded inverter control improved refrigeration efficiency, reduced frost significantly and minimized wasted power during refrigeration. Controller upgraded for easier viewing and operability. Improved temperature distribution accuracy for better incubation. Standard equipped with program operation, auto-stop, auto-start, quick auto stop, timer, calibration off-set, memory, and electricity & CO₂ emission monitor. Designed with analog output (4-20mA) and external communication port (RS485). Operating temperature range 0~ + 60°C.
Catalog No. 50-189-8108 Supplier Yamato Scientific America, Inc. Supplier No. INE800
Add to Cart
Add to Cart
Request a Quote
Programmable , eco-friendly 286L refrigerant incubator. Upgraded inverter control improved refrigeration efficiency, reduced frost significantly and minimized wasted power during refrigeration. Controller upgraded for easier viewing and operability. Improved temperature distribution accuracy for better incubation. Standard equipped with program operation, auto-stop, auto-start, quick auto stop, timer, calibration off-set, memory, and electricity & CO₂ emission monitor. Designed with analog output (4-20mA) and external communication port (RS485). Operating temperature range 0~ + 60°C.