Learn More
Aviva Systems Biology Corp NRG1 Antibody - middle region (ARP46957_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46957_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NRG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NRG1. Alternate names: ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, NDF, SMDF, MST131ApplicationWB. Species reactivity: Cow, Guinea Pig, Human
Catalog No. 50-014-59444