Learn More
Genscript Corporation NRG1 Beta 1 hFc ChimeraBiotin
Catalog No. 5000056723
Encompass
euregulin-1 (NRG-1) is a ligand of the epidermal growth factor receptor (erbB) and its interaction involves activation of the glutamatergic N-methyl-Daspartate receptor which increases the expression of the 2 subunit of the - aminobutyric acid receptor and subunits of the nicotinic acetylcholine receptor
Catalog No. 50-000-56723 Supplier Genscript Corporation Supplier No. Z04575500
euregulin-1 (NRG-1) is a ligand of the epidermal growth factor receptor (erbB) and its interaction involves activation of the glutamatergic N-methyl-Daspartate receptor which increases the expression of the 2 subunit of the - aminobutyric acid receptor and subunits of the nicotinic acetylcholine receptor