Learn More
Medchemexpress LLC NRG1-beta 1 Human 100ug

Supplier: Medchemexpress LLC HYP72007100UG
Recombinant human NRG1-beta 1 (residues S177-E241) is a 65-amino-acid, His-tagged polypeptide produced in E. coli and supplied lyophilized. It is >95% pure and provided with formulation and storage conditions suitable for biochemical and cell-based assays, and is used to study HER2/HER3 signaling and Fn14 regulation in cancer and neurological research.
- Contains 65 amino acids (S177-E241) representing the beta-1 isoform.
- Expressed in E. coli with an N-terminal 6xHis tag for easy purification.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Lyophilized formulation from Tris-NaCl buffer for stability and long-term storage.
- Validated activity in MCF-7 cell proliferation assays (ED50 ~5.7 ng/mL), suitable for signaling studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.