Learn More
Medchemexpress LLC NRG1-beta 1 Human 100ug
SDP
Supplier:  Medchemexpress LLC HYP7365100UG
Encompass_Preferred
NRG1-beta 1 Protein a specific ligand for ERBB3 belongs to a family of structurally related polypeptide growth factors NRG1-beta 1 Protein can regulate cell proliferation differentiation apoptosis and migration NRG1-beta 1 Protein plays an important role in tumors and neurological diseases and also has certain neuroprotective activity NRG-1 Protein Human is a recombinant NRG1-beta 1 protein expressed by E coli without tags[1][2][3][4][5]
Catalog No. 50-001-80014