Learn More
Medchemexpress LLC NRG1-beta 1 Human 100ug
SDP
Catalog No. 5000179233
Encompass_Preferred
HRG1- 1 belongs to a family of structurally-related polypeptide growth factors derived from alternatively spliced genes induces Fn14 expression in MCF7 cells NRG1-beta 1 Protein Human (245a a HEK293 Fc) is the recombinant human-derived NRG1-beta 1 protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-79233 Supplier Medchemexpress LLC Supplier No. HYP73329100UG
HRG1- 1 belongs to a family of structurally-related polypeptide growth factors derived from alternatively spliced genes induces Fn14 expression in MCF7 cells NRG1-beta 1 Protein Human (245a a HEK293 Fc) is the recombinant human-derived NRG1-beta 1 protein expressed by HEK293 with N-hFc labeled tag