Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NRG1-beta 1 Human 10ug
SDP

Catalog No. 5000196451
Encompass_Preferred

Recombinant human neuregulin-1 beta 1 (NRG1-β1) is a bioactive growth factor expressed in CHO cells and provided tag-free as a lyophilized powder. It is designed for cell signaling and neurobiology research, enabling receptor activation and downstream pathway studies with a high-quality, well-characterized ligand.

  • Recombinant human protein expressed in CHO cells.
  • Tag-free formulation to avoid fusion-related artifacts.
  • Lyophilized from 0.22 μm filtered PBS for long-term stability.
  • High purity (>95%) verified by reducing SDS-PAGE.
  • Approximate molecular weight ~7.5 kDa for easy characterization.
  • Supplied in 10 μg vials suitable for assay optimization.

Catalog No. 50-001-96451 Supplier Medchemexpress LLC Supplier No. HYP71171A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human neuregulin-1 beta 1 (NRG1-β1) is a bioactive growth factor expressed in CHO cells and provided tag-free as a lyophilized powder. It is designed for cell signaling and neurobiology research, enabling receptor activation and downstream pathway studies with a high-quality, well-characterized ligand.

  • Recombinant human protein expressed in CHO cells.
  • Tag-free formulation to avoid fusion-related artifacts.
  • Lyophilized from 0.22 μm filtered PBS for long-term stability.
  • High purity (>95%) verified by reducing SDS-PAGE.
  • Approximate molecular weight ~7.5 kDa for easy characterization.
  • Supplied in 10 μg vials suitable for assay optimization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.