Learn More
Medchemexpress LLC NRG1-beta 1 Human 2ug
SDP
Supplier:  Medchemexpress LLC HYP709672UG
Encompass_Preferred
Recombinant human Neuregulin-1 beta 1 is a 246-amino-acid, His-tagged protein expressed in Escherichia coli. It engages ERBB3/ERBB4 receptors to activate downstream signaling pathways and is supplied in purified form for in vitro research applications including receptor signaling studies and cell stimulation assays.
  • Recombinant human protein, 246 amino acids
  • N-terminal 6xHis tag for detection and purification
  • Expressed in Escherichia coli expression system
  • Activates ERBB3/ERBB4 signaling pathways
  • Suitable for cell stimulation and biochemical assays
  • Available in small purified sizes (for example, 2 μg, 10 μg, 50 μg)
Catalog No. 50-001-97175