Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NRG1-beta 1 Human 50ug
SDP

Catalog No. 5000197195
Encompass_Preferred

Recombinant human NRG1-beta 1 produced in CHO cells is a biologically active fragment of neuregulin-1 involved in cell proliferation, differentiation, apoptosis, and neural signaling. Supplied lyophilized and tag-free with a defined amino acid sequence and high purity, it is intended for in vitro research applications in oncology and neuroscience where consistent activity and minimal contaminants are required.

  • High purity (>95%) suitable for sensitive assays.
  • Contains defined amino acid range (T176-K246) for reproducible activity.
  • Tag free to avoid interference in binding studies.
  • Lyophilized formulation for room-temperature shipping and long-term storage.
  • Recommended reconstitution at ≥100 μg/mL and use of carrier protein for extended stability.

Catalog No. 50-001-97195 Supplier Medchemexpress LLC Supplier No. HYP71171A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human NRG1-beta 1 produced in CHO cells is a biologically active fragment of neuregulin-1 involved in cell proliferation, differentiation, apoptosis, and neural signaling. Supplied lyophilized and tag-free with a defined amino acid sequence and high purity, it is intended for in vitro research applications in oncology and neuroscience where consistent activity and minimal contaminants are required.

  • High purity (>95%) suitable for sensitive assays.
  • Contains defined amino acid range (T176-K246) for reproducible activity.
  • Tag free to avoid interference in binding studies.
  • Lyophilized formulation for room-temperature shipping and long-term storage.
  • Recommended reconstitution at ≥100 μg/mL and use of carrier protein for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.