Learn More
Medchemexpress LLC NRG1-beta 2 Human 100ug
SDP
Catalog No. 5000180446
Encompass_Preferred
HRG1- 1 belongs to a family of structurally-related polypeptide growth factors derived from alternatively spliced genes induces Fn14 expression in MCF7 cells NRG1-beta 2 Protein Human is the recombinant human-derived NRG1-beta 2 protein expressed by E coli with tag free
Catalog No. 50-001-80446 Supplier Medchemexpress LLC Supplier No. HYP73304100UG
Add to Cart
Add to Cart
HRG1- 1 belongs to a family of structurally-related polypeptide growth factors derived from alternatively spliced genes induces Fn14 expression in MCF7 cells NRG1-beta 2 Protein Human is the recombinant human-derived NRG1-beta 2 protein expressed by E coli with tag free