Learn More
Aviva Systems Biology Corp NRG3 Antibody - middle region (ARP44588_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44588_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NRG3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NRG3. Alternate names: HRG3, pro-NRG3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58853