Learn More
Aviva Systems Biology Corp Nrip2 Antibody - N-terminal region (ARP37305_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37305_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nrip2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of mouse Nrip2. Alternate names: AW491344, MGC144354, NIX1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-614