Learn More
Aviva Systems Biology Corp NRIP3 Antibody - middle region (ARP50643_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50643_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NRIP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NRIP3. Alternate names: C11orf14, NY-SAR-105ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60399