Learn More
Cayman Chemical nrIum IndIcum ndrosIn 10mg
SDP
Catalog No. 502912869
Encompass_Preferred
A phenol glucoside with diverse biological activities; active against the bacteria S. aureus, S. epidermidis, P. aeruginosa, E. cloacae, K. pneumoniae, and E. coli (MICs = 0.6-1.08 mg/ml) and the fungi C. albicans, C. tropicalis, and C. glabrata (MICs = 1.85, 1.7, and 1.65 mg/ml, respectively); inhibits bronchial obstruction in guinea pig model of asthma induced by ovalbumin or PAF at 10 mg/kg
Catalog No. 50-291-2869 Supplier Cayman Chemical Supplier No. 3619110 MG
A phenol glucoside with diverse biological activities; active against the bacteria S. aureus, S. epidermidis, P. aeruginosa, E. cloacae, K. pneumoniae, and E. coli (MICs = 0.6-1.08 mg/ml) and the fungi C. albicans, C. tropicalis, and C. glabrata (MICs = 1.85, 1.7, and 1.65 mg/ml, respectively); inhibits bronchial obstruction in guinea pig model of asthma induced by ovalbumin or PAF at 10 mg/kg