Learn More
ATCC NRK; Kidney; Rat (Rattus norvegicus)
Catalog No. 502382414
Encompass
NRK is a rat kidney cell line exhibiting epithelial morphology that was isolated from a normal adult rat. This product can be used in cancer and to study renal physiology.
Catalog No. 50-238-2414 Supplier ATCC Supplier No. CRL6509
 Restricted Item
Add to Cart
Add to Cart
NRK is a rat kidney cell line exhibiting epithelial morphology that was isolated from a normal adult rat. This product can be used in cancer and to study renal physiology.