Learn More
AllCells LLC Bone Marrow, Cryopreserved, CD34+ Stem/Progenitor Cells, Positively Selected, 0.5M
Catalog No. 50156615
Encompass
CD34+ Stem/Progenitor cells are positively selected from bone marrow using immunomagnetic CD34+ microbeads. CD34+ cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 90%. Viability is ≥ 80% post-thaw.
Catalog No. 50-156-615 Supplier AllCells LLC Supplier No. BM, CR, CD34+, PS, 0.5M
CD34+ Stem/Progenitor cells are positively selected from bone marrow using immunomagnetic CD34+ microbeads. CD34+ cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 90%. Viability is ≥ 80% post-thaw.