Learn More
AllCells LLC Bone Marrow- Cryopreserved- CD34+ Stem/Progenitor Cells- Positively Selected- 2M
Catalog No. NC2742223
Encompass
CD34+ Stem/Progenitor cells are positively selected from bone marrow using immunomagnetic CD34+ microbeads. CD34+ cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 90%. Viability is ≥ 80% post-thaw.
Catalog No. NC2742223 Supplier AllCells LLC Supplier No. BM CR CD34 PS 2M
CD34+ Stem/Progenitor cells are positively selected from bone marrow using immunomagnetic CD34+ microbeads. CD34+ cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 90%. Viability is ≥ 80% post-thaw.