Learn More
Sino Biological NRP2 / Neuropilin-2 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10695MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NRP2 / Neuropilin-2 (rh NRP2 / Neuropilin-2; Catalog#10695-H08H; NP_003863.2; Met1-Tyr855). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7429