Learn More
Aviva Systems Biology Corp NRTN Antibody - C-terminal region (ARP63641_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63641_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NRTN. It was validated on Western Blot. . Alternate names: NTNApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-288-041