Learn More
Cayman Chemical Nervonic Acid, Purity: ≥95%, CAS: 506-37-6, 100 mg
SDP
Catalog No. 502954544
Encompass_Preferred
A very long chain fatty acid (24:1n-9) enriched in nervous tissue and particularly abundant in sphingolipids, such as sphingomyelin; poorly produced in demyelinating disorders, including multiple sclerosis and adrenoleukodystrophy; binds and inhibits DNA polymerase β (Ki = 4.0 uM) and HIV-1 reverse transcriptase (Ki = 1.2 µM)
Catalog No. 50-295-4544 Supplier Cayman Chemical Supplier No. 13940100 MG
A very long chain fatty acid (24:1n-9) enriched in nervous tissue and particularly abundant in sphingolipids, such as sphingomyelin; poorly produced in demyelinating disorders, including multiple sclerosis and adrenoleukodystrophy; binds and inhibits DNA polymerase β (Ki = 4.0 uM) and HIV-1 reverse transcriptase (Ki = 1.2 µM)