Learn More
Cayman Chemical Nervonic Acid methyl ester, Purity: ≥98%, CAS: 2733-88-2, 50 mg
SDP
Catalog No. 502894236
Encompass_Preferred
An ester version of the free acid which may be more suitable for the formulation of fatty acid-containing diets and dietary supplements
Catalog No. 50-289-4236 Supplier Cayman Chemical Supplier No. 900135150 MG
An ester version of the free acid which may be more suitable for the formulation of fatty acid-containing diets and dietary supplements