Learn More
TARGETMOL CHEMICALS INC NRX-103095 5MG
SDP
Catalog No. 502579408
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NRX-103095 is a potent enhancer of the interaction of (beta)-catenin with the homologous E3 ligase SCF(beta)-TrCP and enhances the affinity of pSer33/Ser37 (beta)-catenin for (beta)-TrCP with an EC50 of 163 nM. purity: 99%
Catalog No. 50-257-9408 Supplier TARGETMOL CHEMICALS INC Supplier No. T637305MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NRX-103095 is a potent enhancer of the interaction of (beta)-catenin with the homologous E3 ligase SCF(beta)-TrCP and enhances the affinity of pSer33/Ser37 (beta)-catenin for (beta)-TrCP with an EC50 of 163 nM. purity: 99%