Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NRX-1532 | 3083833-54-6 | 98.0% | C16H11F3N4O2 | 50 MG
SDP

Catalog No. 5000354506
Encompass_Preferred

NRX-1532 is a small molecule enhancer of β-catenin:β-TrCP interaction. This substance is intended for research use only and is not sold to patients. A publication suggests its function as an enhancer of an E3 ligase-substrate interaction.

  • Small molecule enhancer of β-catenin:β-TrCP interaction
  • Purity: 98.01%
  • Appearance: solid
  • Color: light yellow to yellow
  • Molecular weight: 348.28
  • Chemical formula: C16H11F3N4O2

Catalog No. 50-003-54506 Supplier Medchemexpress LLC Supplier No. HY14498250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NRX-1532 is a small molecule enhancer of β-catenin:β-TrCP interaction. This substance is intended for research use only and is not sold to patients. A publication suggests its function as an enhancer of an E3 ligase-substrate interaction.

  • Small molecule enhancer of β-catenin:β-TrCP interaction
  • Purity: 98.01%
  • Appearance: solid
  • Color: light yellow to yellow
  • Molecular weight: 348.28
  • Chemical formula: C16H11F3N4O2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.