Learn More
Medchemexpress LLC HY-111760 5mg Medchemexpress, NRX-252262 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1117605MG
Encompass_Preferred
Medchemexpress, HY-111760 5mg NRX-252262 CAS: NRX-252262 is a potent enhancer of the interaction between β-Catenin, and its cognate E3 ligase, SCFβ-TrCP, induces mutant β-catenin degradation, with an EC50 of 3.8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8956