Learn More
Medchemexpress LLC NS-102 100mg | 136623-01-3 | 100 MG
SDP
Catalog No. 5000226812
Encompass_Preferred
NS-102 is a selective kainate (GluK2) receptor antagonist and a potent GluR6/7 receptor antagonist used in neuroscience research. It is a small-molecule solid supplied at high purity for in vitro pharmacology and receptor-binding studies.
  • Selective kainate (GluK2) receptor antagonist.
  • Potent GluR6/7 receptor antagonist.
  • CAS 136623-01-3.
  • High purity (≈98%).
  • Solid form suitable for small-scale research preparations.
  • Commonly supplied in milligram-scale quantities (for example, 100 mg).
Catalog No. 50-002-26812 Supplier Medchemexpress LLC Supplier No. HY114427100MG
Add to Cart
Add to Cart
NS-102 is a selective kainate (GluK2) receptor antagonist and a potent GluR6/7 receptor antagonist used in neuroscience research. It is a small-molecule solid supplied at high purity for in vitro pharmacology and receptor-binding studies.
  • Selective kainate (GluK2) receptor antagonist.
  • Potent GluR6/7 receptor antagonist.
  • CAS 136623-01-3.
  • High purity (≈98%).
  • Solid form suitable for small-scale research preparations.
  • Commonly supplied in milligram-scale quantities (for example, 100 mg).