Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NS-102 25mg | 136623-01-3 | 25 MG
SDP

Catalog No. 5000307691
Encompass_Preferred

NS-102 is a selective kainate (GluK2) receptor antagonist and a potent GluR6/7 receptor antagonist supplied as an off-white to yellow solid for research use only. It has molecular formula C12H11N3O4 and a molecular weight of 261.23 g/mol.

  • Selective kainate (GluK2) receptor antagonist for receptor pharmacology research.
  • Off-white to yellow solid form supplied in milligram quantities.
  • Purity approximately 98.5% based on analytical documentation.
  • Molecular formula C12H11N3O4; molecular weight 261.23 g/mol.
  • Store powder at -20°C for long-term stability; in solvent store at -80°C for best stability.

Catalog No. 50-003-07691 Supplier Medchemexpress LLC Supplier No. HY11442725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NS-102 is a selective kainate (GluK2) receptor antagonist and a potent GluR6/7 receptor antagonist supplied as an off-white to yellow solid for research use only. It has molecular formula C12H11N3O4 and a molecular weight of 261.23 g/mol.

  • Selective kainate (GluK2) receptor antagonist for receptor pharmacology research.
  • Off-white to yellow solid form supplied in milligram quantities.
  • Purity approximately 98.5% based on analytical documentation.
  • Molecular formula C12H11N3O4; molecular weight 261.23 g/mol.
  • Store powder at -20°C for long-term stability; in solvent store at -80°C for best stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.