Learn More
Medchemexpress LLC HY-13103 5mg Medchemexpress, NS 11021 CAS:956014-19-0 Purity:>98%
SDP
Catalog No. 501969835
Encompass_Preferred
Medchemexpress, HY-13103 5mg NS 11021 CAS:956014-19-0 NS 11021 is a potent and specific Ca2-activated big-conductance K+ Channels (KCa1.1 channels) activator. NS 11021 at concentrations above 0.3 μM activates KCa1.1 in a concentration-dependent manner by parallelshifting the channel activation curves to more negative potentials. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9835 Supplier Medchemexpress LLC Supplier No. HY131035MG
Medchemexpress, HY-13103 5mg NS 11021 CAS:956014-19-0 NS 11021 is a potent and specific Ca2-activated big-conductance K+ Channels (KCa1.1 channels) activator. NS 11021 at concentrations above 0.3 μM activates KCa1.1 in a concentration-dependent manner by parallelshifting the channel activation curves to more negative potentials. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.