Learn More
Abcam NS 1619, KCa1.1 channel activator, 5MG
Catalog No. 502706417
Encompass_Preferred
MW 362.23 Da, Purity >98%. Potent and selective KCa1.1 channel activator. Induces a concentration-dependent decrease in mitochondrial membrane potential (EC50 = 3.6 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6417 Supplier Abcam Supplier No. AB1418245MG
MW 362.23 Da, Purity >98%. Potent and selective KCa1.1 channel activator. Induces a concentration-dependent decrease in mitochondrial membrane potential (EC50 = 3.6 μM).
The product is subject to the following: Abcam Restricted Use Statement