Learn More
Enzo Life Sciences NS-1643 (10mg). CAS: 448895-37-2
SDP
Catalog No. 502011082
Encompass_Preferred
A novel and selective HERG K+ channel activator (EC50=10.5 µM). Activates ERG2 channels. Displays antiarrhythmic properties in rabbit models. Alternative name: N,N'-Bis[2-hydroxy-5-(trifluoromethyl)phenyl] urea. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or ethanol (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1082 Supplier Enzo Life Sciences Supplier No. BMLKC1690010
A novel and selective HERG K+ channel activator (EC50=10.5 µM). Activates ERG2 channels. Displays antiarrhythmic properties in rabbit models. Alternative name: N,N'-Bis[2-hydroxy-5-(trifluoromethyl)phenyl] urea. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or ethanol (25mg/ml). Long Term Storage: Ambient.