Learn More
Apexbio Technology LLC NS 1643 448895-37-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088823
Encompass_Preferred
NS 1643 (CAS 448895-37-2) is a small-molecule partial agonist of the human hERG potassium channel exhibiting an EC50 of 10 5 M In murine models of triple-negative breast cancer (TNBC) NS 1643 has been shown to inhibit tumor growth via specific signaling pathways Experimental studies demonstrate that NS 1643 suppresses both migration and invasion of breast cancer cells Due to these properties NS 1643 serves as a valuable tool for investigating hERG channel modulation and its impact on cancer cell behavior with potential applications in preclinical oncology research
Catalog No. 50-000-88823 Supplier Apexbio Technology LLC Supplier No. B72605.1
NS 1643 (CAS 448895-37-2) is a small-molecule partial agonist of the human hERG potassium channel exhibiting an EC50 of 10 5 M In murine models of triple-negative breast cancer (TNBC) NS 1643 has been shown to inhibit tumor growth via specific signaling pathways Experimental studies demonstrate that NS 1643 suppresses both migration and invasion of breast cancer cells Due to these properties NS 1643 serves as a valuable tool for investigating hERG channel modulation and its impact on cancer cell behavior with potential applications in preclinical oncology research