Learn More
Enzo Life Sciences NS 1738 (50mg). CAS: 501684-93-1
SDP
Catalog No. 502013400
Encompass_Preferred
A selective positive allosteric modulator selective for α7 nicotinic acetylcholine receptors (α7 nAChR). NS 1738 exhibits cognitive enhancing effects in vivo. Alternative name: 1-(5-chloro-2-hydroxyphenyl)-3-(2-chloro-5-(trifluoromethyl)phenyl)urea, alpha7 nAChR positive allosteric modulator. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: -80°C.
Catalog No. 50-201-3400 Supplier Enzo Life Sciences Supplier No. ENZCHM1170050
A selective positive allosteric modulator selective for α7 nicotinic acetylcholine receptors (α7 nAChR). NS 1738 exhibits cognitive enhancing effects in vivo. Alternative name: 1-(5-chloro-2-hydroxyphenyl)-3-(2-chloro-5-(trifluoromethyl)phenyl)urea, alpha7 nAChR positive allosteric modulator. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: -80°C.