Learn More
Medchemexpress LLC HY-12151 5mg Medchemexpress, NS 1738 CAS:501684-93-1 Purity:>98%
SDP
Catalog No. 502080970
Encompass_Preferred
Medchemexpress, HY-12151 5mg NS 1738 CAS:501684-93-1 NS 1738 (NSC 213859) is a novel positive allosteric modulator of the α7 nAChR, with respect to positive modulation of α7 nAChR (EC50=3.4 μM in oocyte experiments). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0970 Supplier Medchemexpress LLC Supplier No. HY121515MG
Medchemexpress, HY-12151 5mg NS 1738 CAS:501684-93-1 NS 1738 (NSC 213859) is a novel positive allosteric modulator of the α7 nAChR, with respect to positive modulation of α7 nAChR (EC50=3.4 μM in oocyte experiments). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.