Learn More
Medchemexpress LLC NS-3-008 hydrochloride | 1172854-54-4 | 99.7% | 1 ML
SDP
Catalog No. 5000231139
Encompass_Preferred
NS-3-008 hydrochloride is a small-molecule transcriptional inhibitor of G0/G1 switch 2 (G0s2) with a reported IC50 of 2.25 μM. It is supplied as a concentrate solution for research use and as solids for analytical or in vivo studies. Typical applications include biochemical assays and disease-model investigations such as chronic kidney disease research.
  • Transcriptional inhibitor of G0s2 with IC50 ≈ 2.25 μM.
  • Available as 10 mM solution in DMSO (1 mL) and as solid packs for flexibility in workflows.
  • High purity (99.7%) suitable for biochemical, cellular, and in vivo studies.
  • Off-white to light yellow solid with molecular formula C14H24ClN3 and MW 269.81 g·mol⁻¹.
  • Supplied with characterization data to support reproducibility in research.
Catalog No. 50-002-31139 Supplier Medchemexpress LLC Supplier No. HY12089710MM 1ML
NS-3-008 hydrochloride is a small-molecule transcriptional inhibitor of G0/G1 switch 2 (G0s2) with a reported IC50 of 2.25 μM. It is supplied as a concentrate solution for research use and as solids for analytical or in vivo studies. Typical applications include biochemical assays and disease-model investigations such as chronic kidney disease research.
  • Transcriptional inhibitor of G0s2 with IC50 ≈ 2.25 μM.
  • Available as 10 mM solution in DMSO (1 mL) and as solid packs for flexibility in workflows.
  • High purity (99.7%) suitable for biochemical, cellular, and in vivo studies.
  • Off-white to light yellow solid with molecular formula C14H24ClN3 and MW 269.81 g·mol⁻¹.
  • Supplied with characterization data to support reproducibility in research.