Learn More
Medchemexpress LLC HY-14870 50mg , NS-304 Selexipag;ACT-293987 CAS:475086-01-2 Purity:98%
SDP
Catalog No. 501493771
Encompass_Preferred
Medchemexpress, HY-14870 50mg NS-304 Selexipag;ACT-293987 CAS:475086-01-2 Formula:C26H32N4O4S Prostacyclin receptor Purity:98% NS-304 is an orally available and potent agonist for the Prostacyclin ( PGI 2 ) receptor (IP receptor). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3771 Supplier Medchemexpress LLC Supplier No. HY1487050MG
Medchemexpress, HY-14870 50mg NS-304 Selexipag;ACT-293987 CAS:475086-01-2 Formula:C26H32N4O4S Prostacyclin receptor Purity:98% NS-304 is an orally available and potent agonist for the Prostacyclin ( PGI 2 ) receptor (IP receptor). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.