Learn More
Enzo Life Sciences NS-398 (25mg). CAS: 123653-11-2
SDP
Supplier:  Enzo Life Sciences BMLEI2610025
Encompass_Preferred
Cyclooxygenase inhibitor. Selectively inhibits PGH synthase-2 (IC50=1 µM) with no inhibition of PGH synthase-1 at concentrations up to 100 µM. Protects neuronal cultures from LPS-induced neurotoxicity. Alternative name: N-[2-(Cyclohexyloxy)-4-nitrophenyl]-methanesulfonamide. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol (warm; sonication), DMSO (25mg/ml) or dimethyl formamide. Insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-0642