Learn More
TARGETMOL CHEMICALS INC NS-398 50MG
SDP
Catalog No. 502580336
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NS-398 (N-(2-cyclohexyloxy-4-nitrophenyl)methane sulfonamide) is a COX-2 inhibitor wih anti-inflammatory activity. purity: 99%
Catalog No. 50-258-0336 Supplier TARGETMOL CHEMICALS INC Supplier No. T321450MG
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NS-398 (N-(2-cyclohexyloxy-4-nitrophenyl)methane sulfonamide) is a COX-2 inhibitor wih anti-inflammatory activity. purity: 99%