Learn More
Medchemexpress LLC HY-13913 5mg Medchemexpress, NS-398 CAS:123653-11-2 Purity:>98%
SDP
Catalog No. 502081528
Encompass_Preferred
Medchemexpress, HY-13913 5mg NS-398 CAS:123653-11-2 NS-398 is a non-steroidal an-inflammatory agent with analgesic and antipyretic effects, and selectively inhibits prostaglandin G/H synthase 2/cyclooxygenase 2 (COX-2) activity, with an IC50 of 3.8 μM, and has no effect on COX-1 at 100 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1528 Supplier Medchemexpress LLC Supplier No. HY139135MG
Medchemexpress, HY-13913 5mg NS-398 CAS:123653-11-2 NS-398 is a non-steroidal an-inflammatory agent with analgesic and antipyretic effects, and selectively inhibits prostaglandin G/H synthase 2/cyclooxygenase 2 (COX-2) activity, with an IC50 of 3.8 μM, and has no effect on COX-1 at 100 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.