Learn More
Apexbio Technology LLC NS-6180-5MG
SDP
Catalog No. 5000568423
Encompass_Preferred
NS 6180 (CAS No 353262-04-1) is a research compound primarily used in biomedical studies originating from the pharmacological modulation of ion channels It acts by targeting specific potassium channels influencing cellular excitability and maintaining ionic balance This compound has demonstrated activity typically in the nanomolar to low micromolar range in vitro showing significant effects in modulating electrophysiological properties of neurons and other cell types NS 6180 is commonly utilized in cell models and animal studies for understanding neurological functions and disorders It is also deployed in drug screening approaches to identify potential therapeutics Experimental concentrations typically depend on the specific research design ensuring tailored applications in various study protocols
Catalog No. 50-005-68423 Supplier Apexbio Technology LLC Supplier No. B57405
NS 6180 (CAS No 353262-04-1) is a research compound primarily used in biomedical studies originating from the pharmacological modulation of ion channels It acts by targeting specific potassium channels influencing cellular excitability and maintaining ionic balance This compound has demonstrated activity typically in the nanomolar to low micromolar range in vitro showing significant effects in modulating electrophysiological properties of neurons and other cell types NS 6180 is commonly utilized in cell models and animal studies for understanding neurological functions and disorders It is also deployed in drug screening approaches to identify potential therapeutics Experimental concentrations typically depend on the specific research design ensuring tailored applications in various study protocols