Learn More
Medchemexpress LLC NS-638 | 150493-34-8 | 99.8% | 325.72 g/mol | C15H11ClF3N3 | 2 MG
SDP
Catalog No. 5000247655
Encompass_Preferred
NS-638 is a small nonpeptide research compound that acts as a Ca2+-channel blocker. It inhibits K+-stimulated intracellular Ca2+ elevation with an IC50 of 3.4 μM and is supplied as a solid or as DMSO solutions for laboratory use.
  • Potent Ca2+ channel blocker with IC50 3.4 μM.
  • High supplied purity (reported 99.84%).
  • Available as solid and 10 mM DMSO solution formats.
  • Molecular weight 325.72 g/mol.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (2 years) or -20°C (1 year).
Catalog No. 50-002-47655 Supplier Medchemexpress LLC Supplier No. HY1014282MG
NS-638 is a small nonpeptide research compound that acts as a Ca2+-channel blocker. It inhibits K+-stimulated intracellular Ca2+ elevation with an IC50 of 3.4 μM and is supplied as a solid or as DMSO solutions for laboratory use.
  • Potent Ca2+ channel blocker with IC50 3.4 μM.
  • High supplied purity (reported 99.84%).
  • Available as solid and 10 mM DMSO solution formats.
  • Molecular weight 325.72 g/mol.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (2 years) or -20°C (1 year).