Learn More
Medchemexpress LLC NS-638 | 150493-34-8 | 99.8% | 325.72 g/mol | C15H11ClF3N3 | 50 MG
SDP
Catalog No. 5000249143
Encompass_Preferred
NS-638 is a small nonpeptide calcium channel blocker used in research to inhibit K+-stimulated intracellular Ca2+ elevation (IC50 = 3.4 μM). It is supplied with high reported purity and is available as a powder or ready-to-use DMSO solutions for use in biochemical and cellular assays.
  • High purity suitable for biochemical and cellular assays.
  • Blocks K+-stimulated intracellular Ca2+ elevation (IC50 3.4 μM).
  • Available as powder and as DMSO solution for easy reconstitution.
  • Provides long-term stability when stored under recommended conditions.
  • Intended for in vitro pharmacology and calcium channel studies.
Catalog No. 50-002-49143 Supplier Medchemexpress LLC Supplier No. HY10142850MG
NS-638 is a small nonpeptide calcium channel blocker used in research to inhibit K+-stimulated intracellular Ca2+ elevation (IC50 = 3.4 μM). It is supplied with high reported purity and is available as a powder or ready-to-use DMSO solutions for use in biochemical and cellular assays.
  • High purity suitable for biochemical and cellular assays.
  • Blocks K+-stimulated intracellular Ca2+ elevation (IC50 3.4 μM).
  • Available as powder and as DMSO solution for easy reconstitution.
  • Provides long-term stability when stored under recommended conditions.
  • Intended for in vitro pharmacology and calcium channel studies.